Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAMTS15 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5149U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5149U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5149U100 Supplier R&D Systems Supplier No. FAB5149U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAMTS15 Monoclonal specifically detects ADAMTS15 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ADAMTS15
Applications Flow Cytometry
Classification Monoclonal
Clone 561819
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8TE58
Gene Alias A disintegrin and metalloproteinase with thrombospondin motifs 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, preproprotein, ADAM metallopeptidase with thrombospondin type 1 motif, 15, ADAM-TS 15, ADAM-TS15, ADAMTS-15, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, metalloprotease disintegrin 15 with thrombospondin domains, MGC126403
Gene Symbols ADAMTS15
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS15 Gly18-Cys682 Accession # Q8TE58
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170689
Test Specificity Detects human ADAMTS15 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.