Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ADAMTS4 MAb (Clone 416610), R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4307
This item is not returnable.
View return policy
Description
ADAMTS4 Monoclonal specifically detects ADAMTS4 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry.Specifications
ADAMTS4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS2, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1, ADMP-1EC 3.4.24.82, Aggrecanase 1, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9507 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunoprecipitation, Neutralization, Immunocytochemistry | |
416610 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
O75173 | |
ADAMTS4 | |
Mouse myeloma cell line NS0-derived recombinant human ADAMTS4 Phe213-Cys685 Accession # O75173 | |
500 μg | |
Primary | |
Detects human ADAMTS4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ADAMTS1, rhADAMTS5, rhADAMTS13, or rhADAMTSL1.2 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction