Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAMTS5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2198
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2198 100 μg
AF2198SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2198 Supplier R&D Systems Supplier No. AF2198
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ADAMTS5 Polyclonal specifically detects ADAMTS5 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ADAMTS5
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q9UNA0
Gene Alias A disintegrin and metalloproteinase with thrombospondin motifs 11, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 5 (aggrecanase-2), ADAM metallopeptidase with thrombospondin type 1 motif, 5, ADAM-TS 11, ADAM-TS 5, ADAMTS11, ADAMTS-11, ADAM-TS5, ADAMTS-5, ADMP2, ADMP-2, ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5, Aggrecanase 2, Aggrecanase-2, disintegrin-like and metalloprotease with thrombospondin type 1 motif, 510ADAMTS11FLJ36738, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82
Gene Symbols ADAMTS5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ADAMTS5 Ser262-Pro622 Accession # Q9UNA0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11096
Test Specificity Detects human ADAMTS5 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) ADAMTS1 and rhADAMTS-L1.2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.