Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAMTSL-1/Punctin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1717SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1717SP 25 μg
MAB1717 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1717SP Supplier R&D Systems Supplier No. MAB1717SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAMTSL-1/Punctin Monoclonal specifically detects ADAMTSL-1/Punctin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ADAMTSL-1/Punctin
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 212125
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8N6G6
Gene Alias ADAM-TS related protein 1, ADAMTSL1, ADAMTSL-1, ADAMTS-like 1, ADAMTS-like protein 1, ADAMTSR1MGC118805, C9orf94, chromosome 9 open reading frame 94, DKFZp686L03130, FLJ35283, FLJ41032, FLJ46891, MGC118803, MGC40193, PUNCTIN, Punctin-1
Gene Symbols ADAMTSL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ADAMTSL-1/Punctin Glu29-Ser525 Accession # Q8N6G6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92949
Test Specificity Detects human ADAMTSL-1/Punctin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.