Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adenosine A1R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11135SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11135SP 25 μg
MAB11135100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11135SP Supplier R&D Systems Supplier No. MAB11135SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenosine A1R Monoclonal antibody specifically detects Adenosine A1R in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Adenosine A1R
Applications Immunohistochemistry
Classification Monoclonal
Clone 1051024
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P30542
Gene Alias Adenosine A1 R, adenosine A1 receptor, ADORA 1, ADORA1, RDC7, RDC7adenosine receptor A1
Host Species Mouse
Immunogen Human Adora-1 synthetic peptide corresponding to the 2nd. extra-cellular loop.,, Accession # P30542
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.