Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Adenosine A2b R Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB104442U100UG
This item is not returnable.
View return policy
Description
Adenosine A2b R Monoclonal specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry.Specifications
Adenosine A2b R | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
adenosine A2b receptor, Adenosine A2bR, adenosine receptor A2b, ADORA2, ADORA2B | |
Human Adenosine A2b R synthetic peptide, Accession # NP_000667 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Flow Cytometry | |
2683F | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_000667 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
136 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction