Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adenosine A2b R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB104442SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB104442SP 25 μg
MAB10444210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB104442SP Supplier R&D Systems Supplier No. MAB104442SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Adenosine A2b R Monoclonal antibody specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Adenosine A2b R
Applications Flow Cytometry
Classification Monoclonal
Clone 2683F
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/106 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_000667
Gene Alias adenosine A2b receptor, Adenosine A2bR, adenosine receptor A2b, ADORA2, ADORA2B
Host Species Rabbit
Immunogen Human Adenosine A2b R synthetic peptide, Accession # NP_000667
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.