Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adiponectin/Acrp30 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1065
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1065 Supplier R&D Systems Supplier No. AF1065
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Adiponectin/Acrp30 Polyclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Adiponectin/Acrp30
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Accession No. Q15848
Gene Alias ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein
Gene Symbols ADIPOQ
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Adiponectin/Acrp30 Glu19-Asn244 Accession # Q15848
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Test Specificity Detects human Adiponectin/Acrp30 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Adiponectin is observed and less than 5% cross-reactivity with recombinant rat Adiponectin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.