Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Adiponutrin/PNPLA3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$177.10 - $443.50
Specifications
Antigen | Adiponutrin/PNPLA3 |
---|---|
Clone | 847516 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5208100
![]() |
R&D Systems
MAB5208100 |
100 μg |
Each for $443.50
|
|
|||||
MAB5208SP
![]() |
R&D Systems
MAB5208SP |
25 μg |
Each for $177.10
|
|
|||||
Description
Adiponutrin/PNPLA3 Monoclonal antibody specifically detects Adiponutrin/PNPLA3 in Human samples. It is validated for ImmunohistochemistrySpecifications
Adiponutrin/PNPLA3 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Acylglycerol O-acyltransferase, adiponutrin, ADPN, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3, PNPLA3 | |
E. coli -derived recombinant human Adiponutrin/PNPLA3, Arg160-Arg349, Accession # Q9NST1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
847516 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9NST1 | |
80339 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title