Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adiponutrin/PNPLA3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5208
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5208 Supplier R&D Systems Supplier No. AF5208
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Adiponutrin/PNPLA3 Polyclonal specifically detects Adiponutrin/PNPLA3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Adiponutrin/PNPLA3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9NST1
Gene Alias Acylglycerol O-acyltransferase, adiponutrin, ADPN, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3, PNPLA3
Gene Symbols PNPLA3
Host Species Sheep
Immunogen E. coli-derived recombinant human Adiponutrin/PNPLA3 Arg160-Arg349 Accession # Q9NST1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80339
Test Specificity Detects human Adiponutrin/PNPLA3 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.