Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AdipoR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10128R100UG
This item is not returnable.
View return policy
Description
AdipoR1 Monoclonal specifically detects AdipoR1 in Human samples. It is validated for Flow Cytometry.Specifications
AdipoR1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human AdipoR1 in direct ELISAs. In direct ELISA, no cross-reactivity with human AdipoR2 is observed | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
2416C | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
ACDCR1, adiponectin receptor 1, adiponectin receptor protein 1, ADR1, CGI45, FLJ42464, PAQR1, PAQR1FLJ25385, Progestin and adipoQ receptor family member I, TESBP1A | |
Synthetic peptide containing human AdipoR1 C-term sequences, | |
100 μg | |
51094.0 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction