Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Adrenomedullin/ADM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB61081100
This item is not returnable.
View return policy
Description
Adrenomedullin/ADM Monoclonal specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry.Specifications
Adrenomedullin/ADM | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ADM, adrenomedullin, AMpreproadrenomedullin | |
Human Adrenomedullin/ADM synthetic peptide, Accession # P35318 | |
100 μg | |
133 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Immunohistochemistry | |
1003019 | |
Immunohistochemistry 5-25 μg/mL | |
P35318 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Adrenomedullin/ADM in direct ELISAs. | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction