Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adrenomedullin R/ADMR Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10293T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10293T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10293T10 Supplier R&D Systems Supplier No. FAB10293T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adrenomedullin R/ADMR/GPR182 Monoclonal specifically detects Adrenomedullin R/ADMR/GPR182 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Adrenomedullin R/ADMR/GPR182
Applications Flow Cytometry
Classification Monoclonal
Clone 528563
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O15218
Gene Alias 7TMR, ADMR, adrenomedullin receptor, AdrenomedullinR, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, GAMRH, Gpcr22, GPR182, G-protein coupled receptor 182, hrhAMR, MB10, MGC34399, NOW
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Adrenomedullin R/ADMR, Accession # O15218
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11318
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.