Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Adrenomedullin R/ADMR Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $456.00

Specifications

Antigen Adrenomedullin R/ADMR/GPR182
Clone 528563
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10293SP
GREENER_CHOICE
View Documents
R&D Systems
MAB10293SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB10293100
GREENER_CHOICE
View Documents
R&D Systems
MAB10293100
100 μg
Each for $456.00
Only null left
Add to Cart
 
Description

Description

Adrenomedullin R/ADMR/GPR182 Monoclonal specifically detects Adrenomedullin R/ADMR/GPR182 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications

Specifications

Adrenomedullin R/ADMR/GPR182
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
7TMR, ADMR, adrenomedullin receptor, AdrenomedullinR, AMR, AM-R, G protein-coupled receptor 182,7TMR, G10D, GAMRH, Gpcr22, GPR182, G-protein coupled receptor 182, hrhAMR, MB10, MGC34399, NOW
HEK293 human embryonic kidney cell line transfected with human Adrenomedullin R/ADMR, Accession # O15218
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Adrenomedullin R/ADMR in direct ELISAs.
528563
Flow Cytometry, Immunohistochemistry, CyTOF
Unconjugated
Mouse
Human
O15218
11318
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.