Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Afamin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8065SP
This item is not returnable.
View return policy
Description
Afamin Polyclonal specifically detects Afamin in Human samples. It is validated for Western Blot.Specifications
Afamin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
afamin, AFM, ALB2, ALB2alpha-Alb, ALBA, ALBAalpha-albumin, ALF, Alpha-Alb, Alpha-albumin, MGC125338, MGC125339 | |
Sheep | |
Affinity Purified | |
RUO | |
173 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
P43652 | |
AFM | |
Human embryonic kidney cell line HEK293-derived recombinant human Afamin Leu22-Asn599 Accession # P43652 | |
25 μg | |
Primary | |
Detects human Afamin in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Afamin is observed, and less than 1% cross-reactivity with recombinant human (rh) AFB and rhAFP is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction