Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Afamin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8065
This item is not returnable.
View return policy
Description
Afamin Polyclonal specifically detects Afamin in Human samples. It is validated for Western Blot.Specifications
| Afamin | |
| Polyclonal | |
| Unconjugated | |
| P43652 | |
| AFM | |
| Human embryonic kidney cell line HEK293-derived recombinant human Afamin Leu22-Asn599 Accession # P43652 | |
| 100 μg | |
| Primary | |
| Detects human Afamin in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Afamin is observed, and less than 1% cross-reactivity with recombinant human (rh) AFB and rhAFP is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| afamin, AFM, ALB2, ALB2alpha-Alb, ALBA, ALBAalpha-albumin, ALF, Alpha-Alb, Alpha-albumin, MGC125338, MGC125339 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 173 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction