Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aggrecan (ADAMTS5-cleaved) Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Aggrecan |
---|---|
Clone | 661725 |
Concentration | LYOPH |
Dilution | Immunoprecipitation 25 ug/mL |
Applications | Immunoprecipitation |
Description
Aggrecan Monoclonal specifically detects Aggrecan in Human samples. It is validated for Immunoprecipitation.Specifications
Aggrecan | |
LYOPH | |
Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P16112 | |
176 | |
KLHG-conjugated peptide from human Aggrecan, Leu384-Glu392 Accession # P16112 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human Aggrecan in direct ELISAs. It specifically recognizes the N-terminal fragment of recombinant human Aggrecan released following cleavage by ADAMTS5. |
661725 | |
Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK | |
ACAN | |
IgG3 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title