Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aggrecan (ADAMTS5-cleaved) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB64891
This item is not returnable.
View return policy
Description
Aggrecan Monoclonal specifically detects Aggrecan in Human samples. It is validated for Western Blot.Specifications
Aggrecan | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P16112 | |
ACAN | |
Aggrecan cleaved C-term aa 393-402 ARGSVILTVK Accession # P16112 | |
100 μg | |
Primary | |
Detects human Aggrecan in direct ELISAs and Western blots. It specifically recognizes the C-terminal fragment of recombinant human Aggrecan released following cleavage by ADAMTS5. | |
Human | |
Purified |
Western Blot | |
726006 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
176 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction