Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Aggrecan G1-IGD-G2 Domains Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1220SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1220SP 25 μg
MAB1220 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1220SP Supplier R&D Systems Supplier No. MAB1220SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Aggrecan Monoclonal specifically detects Aggrecan in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Aggrecan
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 179509
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL, Immunoprecipitation 25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_037359
Gene Alias ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK
Gene Symbols ACAN
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Aggrecan Val20-Gly675 Accession # NP_037359
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 176
Test Specificity Detects human Aggrecan G1-IGD-G2 Domains in Western blots and direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.