Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AGTR-1 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10244U10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10244U10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10244U10 Supplier R&D Systems Supplier No. FAB10244U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AGTR-1 Monoclonal specifically detects AGTR-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen AGTR-1
Applications Flow Cytometry
Classification Monoclonal
Clone 1010103
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias AG2S, AGTR1, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1, AT1AR, AT1AT1AR, AT1B, AT1BR, AT1R, AT2R1, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor
Host Species Mouse
Immunogen Synthetic peptide containing human AGTR-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 185
Test Specificity Detects human AGTR-1 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.