Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AGTR-1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10244R100UG
This item is not returnable.
View return policy
Description
AGTR-1 Monoclonal specifically detects AGTR-1 in Human samples. It is validated for Flow Cytometry.Specifications
AGTR-1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human AGTR-1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Flow Cytometry | |
1010103 | |
Flow Cytometry 0.25-1 μg/106 cells | |
AG2S, AGTR1, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1, AT1AR, AT1AT1AR, AT1B, AT1BR, AT1R, AT2R1, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor | |
Synthetic peptide containing human AGTR-1 | |
100 μg | |
185 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction