Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AICDA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB39102
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB39102 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB39102 Supplier R&D Systems Supplier No. MAB39102
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

AICDA Monoclonal specifically detects AICDA in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen AICDA
Applications Immunocytochemistry
Classification Monoclonal
Clone 359218
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9GZX7
Gene Alias activation-induced cytidine deaminase, AID, AIDCytidine aminohydrolase, Arp2, CDA2, HIGM2, HIGM2EC 3.5.4.5, integrated into Burkitt's lymphoma cell line Ramos
Gene Symbols AICDA
Host Species Rat
Immunogen E. coli-derived recombinant human AICDA Met1-Leu198 Accession # Q9GZX7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57379
Test Specificity Detects human AICDA in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.