Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AICL/CLEC-2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$196.30 - $484.50
Specifications
Antigen | AICL/CLEC-2B |
---|---|
Clone | 1064941 |
Dilution | Western Blot 2 μg/mL, Immunohistochemistry 5 μg/mL, Immunocytochemistry 8 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB90591100
![]() |
R&D Systems
MAB90591100 |
100 μg |
Each for $484.50
|
|
|||||
MAB90591SP
![]() |
R&D Systems
MAB90591SP |
25 μg |
Each for $196.30
|
|
|||||
Description
AICL/CLEC-2B Monoclonal antibody specifically detects AICL/CLEC-2B in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunocytochemistrySpecifications
AICL/CLEC-2B | |
Western Blot 2 μg/mL, Immunohistochemistry 5 μg/mL, Immunocytochemistry 8 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Activation-induced C-type lectin, AICLC-type lectin superfamily member 2, CLEC2B, CLEC-2B, CLECSF2, CLECSF2IFN-alpha-2b-inducing-related protein 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 2 (activation-induced), C-type lectin domain family 2 member B, C-type lectin domain family 2, member B, HP10085, IFN-alpha2b-inducing related protein 1, IFNRG1 | |
Chinese Hamster Ovary cell line, CHO-derived human AICL/CLEC-2B, Lys26-His149, Accession # Q92478 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1064941 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q92478 | |
9976 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title