Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AICL/CLEC-2B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9059100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9059100 100 μg
MAB9059SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9059100 Supplier R&D Systems Supplier No. MAB9059100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AICL/CLEC-2B Monoclonal antibody specifically detects AICL/CLEC-2B in Human samples. It is validated for Flow Cytometry

Specifications

Antigen AICL/CLEC-2B
Applications Flow Cytometry
Classification Monoclonal
Clone 1064935
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q92478
Gene Alias Activation-induced C-type lectin, AICLC-type lectin superfamily member 2, CLEC2B, CLEC-2B, CLECSF2, CLECSF2IFN-alpha-2b-inducing-related protein 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 2 (activation-induced), C-type lectin domain family 2 member B, C-type lectin domain family 2, member B, HP10085, IFN-alpha2b-inducing related protein 1, IFNRG1
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human AICL/CLEC-2B, Lys26-His149, Accession # Q92478
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9976
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.