Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human AIM2 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266677
Click to view available options
Quantity:
1 Kit

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human AIM2 ELISA Kit (Chemiluminescence) measures human AIM2 in serum, plasma and other biological fluids.

Specifications

Assay Range 31.25-2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Cross Reactivity This kit recognizes Human AIM2 in samples. No significant cross-reactivity or interference between Human AIM2 and analogues was observed.
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 9447
Gene Symbol AIM2
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target AIM2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.