Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Aiolos/IKZF3 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  R&D Systems FAB8625S100UG

Catalog No. FAB8625S100


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Aiolos/IKZF3 Monoclonal specifically detects Aiolos/IKZF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications

Specifications

Aiolos/IKZF3
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q9UKT9
IKZF3
E. coli-derived recombinant human Aiolos/IKZF3 Met1-Gly54 Accession # Q9UKT9
100 μg
Primary
Detects human Aiolos/IKZF3 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
914821
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, IKZF3, zinc finger protein Aiolos, ZNFN1A3, ZNFN1A3subfamily 1A, 3 (Aiolos)
Mouse
Protein A or G purified from cell culture supernatant
RUO
22806
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only (RUO)