Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Airway Trypsin-like Protease/HAT Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2695
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2695 Supplier R&D Systems Supplier No. MAB2695
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Airway Trypsin-like Protease/HAT/TMPRSS11D Monoclonal specifically detects Airway Trypsin-like Protease/HAT/TMPRSS11D in Human samples. It is validated for Western Blot.

Specifications

Antigen Airway Trypsin-like Protease/HAT/TMPRSS11D
Applications Western Blot
Classification Monoclonal
Clone 337029
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O60235
Gene Alias airway trypsin like protease, Airway trypsin-like protease, EC 3.4.21, EC 3.4.21.-, HAT, MGC150587, MGC150588, TMPRSS11D, transmembrane protease serine 11D, transmembrane protease, serine 11D
Gene Symbols TMPRSS11D
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Airway Trypsin-like Protease/HAT Ala42-Ile418 Accession # O60235
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9407
Test Specificity Detects human Airway Trypsin-like Protease/HAT in direct ELISAs and Western blots. Recognizes the catalytic-domain of recombinant human HAT.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.