Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Akt Pan Specific Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF2055
This item is not returnable.
View return policy
Description
AKT Polyclonal specifically detects AKT in Human samples. It is validated for Western Blot, Simple Western.Specifications
AKT | |
Polyclonal | |
Unconjugated | |
P31749 | |
AKT1 | |
Akt synthetic peptide IRCLQWTTVIERTFHV Accession # P31749 | |
50 μg | |
Primary | |
Detects human Akt1, Akt2, and Akt3 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 20 ug/mL | |
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma | |
Rabbit | |
Affinity Purified | |
RUO | |
207 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction