Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Akt2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC23152N100UG
This item is not returnable.
View return policy
Description
AKT2 Monoclonal specifically detects AKT2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
AKT2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P31751 | |
AKT2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt2 Asn2-Glu481 Accession # P31751 | |
RUO | |
208 | |
Human | |
IgG1 |
Flow Cytometry | |
302501 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
Mouse | |
100 μg | |
Primary | |
Detects human Akt2. Using direct ELISA, this antibody does not detect recombinant human (rh) Akt1 or rhAkt3. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction