Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Akt3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC14631V100UG
This item is not returnable.
View return policy
Description
AKT3 Monoclonal specifically detects AKT3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
AKT3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_005456 | |
AKT3 | |
E. coli-derived recombinant human Akt3 Arg23-Ala101 Accession # NP_005456 | |
RUO | |
10000 | |
Human | |
IgG2a |
Flow Cytometry | |
199822 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, RAC-gamma, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma) | |
Mouse | |
100 μg | |
Primary | |
Detects human Akt3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Akt1 or rhAkt2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction