Learn More
Human ALCAM/CD166 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ALCAM/CD166 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2599A |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q13740 |
| Gene Alias | activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.