Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ALCAM/CD166 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF656
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF656 100 μg
AF656SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF656 Supplier R&D Systems Supplier No. AF656
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

ALCAM/CD166 Polyclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, Adhesion Blockade.

Specifications

Antigen ALCAM/CD166
Applications Western Blot, Adhesion Blockade
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Adhesion Blockade
Gene Accession No. AAB59499
Gene Alias activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Gene Symbols ALCAM
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # AAB59499
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214
Test Specificity Detects human ALCAM/CD166 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse ALCAM/CD166 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.