Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aldo-keto Reductase 1C3/AKR1C3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7678
This item is not returnable.
View return policy
Description
Aldo-keto Reductase 1C3/AKR1C3 Polyclonal specifically detects Aldo-keto Reductase 1C3/AKR1C3 in Human samples. It is validated for Western Blot.Specifications
Aldo-keto Reductase 1C3/AKR1C3 | |
Polyclonal | |
Unconjugated | |
P42330 | |
AKR1C3 | |
E. coli-derived recombinant human Aldo-keto Reductase 1C3/AKR1C3 Met1-Tyr323 Accession # P42330 | |
100 μg | |
Primary | |
Detects human Aldo-keto Reductase 1C3/AKR1C3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) AKR1C1 is observed, and approximately 25% cross-reactivity with rhAKR1C4 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
17-beta-hydroxysteroid dehydrogenase type 5, 3-alpha-HSD type 2, 3-alpha-hydroxysteroid dehydrogenase type 2, AKR1C3, aldo-keto reductase family 1 member C3, aldo-keto reductase family 1, member C3 (3-alpha hydroxysteroid dehydrogenase, type II), AldoketoReductase 1C3, Chlordecone reductase homolog HAKRb, DD3, DD-3, DD317-beta-HSD 5, DDH1, DDX, Dihydrodiol dehydrogenase 3, Dihydrodiol dehydrogenase type I, dihydrodiol dehydrogenase X, EC 1.1.1, EC 1.1.1.112,3-alpha-HSD type II, brain, EC 1.1.1.188, EC 1.1.1.213, EC 1.1.1.63, EC 1.1.1.64, EC 1.3.1.20, HA1753, HAKRB, HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase, HAKRe, HSD17B5, hydroxysteroid (17-beta) dehydrogenase 5, Indanol dehydrogenase, KIAA0119hluPGFS, PGFS, Prostaglandin F synthase, Testosterone 17-beta-dehydrogenase 5, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase | |
Sheep | |
Affinity Purified | |
RUO | |
8644 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction