Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ALK-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$227.50 - $596.50
Specifications
Antigen | ALK-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
ALK-1 Polyclonal specifically detects ALK-1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ALK-1 | |
Western Blot 0.1 ug/mL, Immunoprecipitation | |
Polyclonal | |
Goat | |
Human | |
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
ACVRL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P37023 | |
94 | |
Mouse myeloma cell line NS0-derived recombinant human ALK-1 Asp22-Gln118 Accession # P37023 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ALK-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse ALK-1 is observed and less than 1% cross-reactivity with recombinant human (rh) Activin RIA and rhActivin RIB is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title