Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ALK-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB370SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB370SP 25 μg
MAB370100 100 μg
MAB370500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB370SP Supplier R&D Systems Supplier No. MAB370SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen ALK-1
Applications Western Blot
Classification Monoclonal
Clone 117720
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 40 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P37023
Gene Alias activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Gene Symbols ACVRL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ALK-1 Asp22-Gln118 Accession # P37023
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94
Test Specificity Detects human ALK-1 in direct ELISAs and Western blots. Shows less than 1% cross-reactivity with recombinant mouse ALK-1, recombinant human (rh) Activin RIA, or rhActivin RIB.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.