Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human ALK-1 - Ready-To-Use ELISA Kit (Colorimetric)
SDP

Catalog No. NB468658
Click to view available options
Quantity:
1 Kit

Colorimetric ELISA Kit

Specifications

Assay Range 0.156 - 10 ng/mL (example only; lot dependent)
Assay Sensitivity 0.054 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Tissue Homogenates, Cell Lysates, Biological Fluids
For Use With (Application) ELISA
Gene ID (Entrez) 94
Gene Symbol ACVRL1
Quantity 1 Kit
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.