Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ALK-7 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA77491G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA77491G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA77491G100 Supplier R&D Systems Supplier No. FAB77491G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ALK-7/Activin Receptor Type 1C
Applications Flow Cytometry
Classification Monoclonal
Clone 810506
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8NER5
Gene Alias activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30
Gene Symbols ACVR1C
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ALK-7 Leu26-Glu113 Accession # Q8NER5
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130399
Test Specificity Detects human ALK-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat ALK-7 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.