Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Alkaline Phosphatase/ALPL Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2909
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western Blot.Specifications
Alkaline Phosphatase/ALPL | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P05186 | |
ALPL | |
E. coli-derived recombinant human Alkaline Phosphatase/ALPL Leu18-Ser502 Accession # P05186 | |
100 μg | |
Primary | |
Detects human Alkaline Phosphatase/ALPL in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
388816 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
249 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction