Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Alkaline Phosphatase/ ALPL Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM1448
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM1448 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM1448 Supplier R&D Systems Supplier No. BAM1448
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Alkaline Phosphatase/ALPL
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone B4-78
Concentration LYOPH
Conjugate Biotin
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Alias Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935
Gene Symbols ALPL
Host Species Mouse
Immunogen Human liver, bone and kidney-derived Alkaline Phosphatase/ALPL
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 249
Test Specificity Detects liver, bone and kidney Alkaline Phosphatase/ALPL from human tissue (2).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.