Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Alkaline Phosphatase/ALPL PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
$221.50 - $444.00
Specifications
Antigen | Alkaline Phosphatase/ALPL |
---|---|
Clone | B4-78 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1448P025
![]() |
R&D Systems
FAB1448P025 |
25 Tests |
Each for $221.50
|
|
|||||
FAB1448P100
![]() |
R&D Systems
FAB1448P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Flow Cytometry.Specifications
Alkaline Phosphatase/ALPL | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
249 | |
Human liver, bone and kidney-derived Alkaline Phosphatase/ALPL | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
B4-78 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
ALPL | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects liver, bone and kidney Alkaline Phosphatase/ALPL from human tissue (2). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title