Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Alkaline Phosphatase/ALPP/ALPI Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5905
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5905 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5905 Supplier R&D Systems Supplier No. AF5905
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Alkaline Phosphatase/ALPP/ALPI Polyclonal specifically detects Alkaline Phosphatase/ALPP/ALPI in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.

Specifications

Antigen Alkaline Phosphatase/ALPP/ALPI
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P05187
Gene Symbols ALPP
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Alkaline Phosphatase/ALPP Ile23-Asp506 Accession # P05187
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5710
Test Specificity Detects recombinant human Alkaline Phosphatase Placental Type/ALPP and recombinant human Alkaline Phosphatase Intestinal Type/ALPI in direct ELISAs. Detects human ALPP and ALPI but not Alkaline Phosphatase Liver Type/ALPL in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.