Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Allergin-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7364SP
This item is not returnable.
View return policy
Description
Allergin-1 Polyclonal specifically detects Allergin-1 in Human samples. It is validated for Western Blot.Specifications
Allergin-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Allergin1, Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, MCA32-1S2, MILR1, Probable Mast Cell Antigen 32 Homolog | |
Sheep | |
Affinity Purified | |
RUO | |
284021 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
BAJ08253 | |
MILR1 | |
E. coli-derived recombinant human Allergin-1 (isoform Allergin-1S2) Met29-Ser131 Accession # BAJ08253 | |
25 μg | |
Primary | |
Detects human Allergin-1 in direct ELISAs. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction