Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-1 Adrenergic R/ADRA1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10298SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10298SP 25 μg
MAB10298100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10298SP Supplier R&D Systems Supplier No. MAB10298SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-1A Adrenergic R/ADRA1A Monoclonal specifically detects alpha-1A Adrenergic R/ADRA1A in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen alpha-1A Adrenergic R/ADRA1A
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 518508
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. P35348
Gene Alias a-1A AdrenergicR, ADRA1A, ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, alpha1A AdrenergicR, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human alpha-1 Adrenergic R/ADRA1A, Accession # P35348
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.