Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-2B Adrenergic R/ADRA2B Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10324S100UG
This item is not returnable.
View return policy
Description
alpha-2B Adrenergic R/ADRA2B Monoclonal specifically detects alpha-2B Adrenergic R/ADRA2B in Human samples. It is validated for Flow Cytometry.Specifications
alpha-2B Adrenergic R/ADRA2B | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
a-2B AdrenergicR, ADRA2B, ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, alpha2B AdrenergicR, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor | |
NS0 mouse myeloma cell line transfected with human alpha-2B Adrenergic R/ADRA2B, Accession # P18089 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
491613 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P18089 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
151 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction