Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-Defensin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8198
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8198 Supplier R&D Systems Supplier No. AF8198
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

alpha-Defensin 1 Polyclonal specifically detects alpha-Defensin 1 in Human samples. It is validated for ELISA, Immunohistochemistry.

Order Info

The product is shipped at ambient temperature.

Specifications

Antigen alpha-Defensin 1
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P59665
Gene Alias alpha 2, alphaDefensin 1, DEF1, DEF1DEFA1B, DEFA1, DEFA2, defensin, alpha 1, myeloid-related sequence, defensin, alpha 1HP-1, HNP-1, HNP-1HP1, HP1, MRS, MRSMGC138393, myeloid-related sequence, neutrophil defensin 1
Gene Symbols DEFA1
Host Species Sheep
Immunogen E. coli-derived recombinant human alpha-Defensin 1 Ala65-Cys94 Accession # P59665
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1667
Test Specificity Detects human alpha-Defensin 1 in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.