Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-Defensin 5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB102481SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB102481SP 25 μg
MAB10248110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB102481SP Supplier R&D Systems Supplier No. MAB102481SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for ELISA.

Specifications

Antigen Defensin alpha 5
Applications ELISA
Classification Monoclonal
Clone 972217
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q01523
Gene Alias DEF5, DEF5defensin, alpha 5, preproprotein, DEFA5, defensin 5, Defensin alpha 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha-Defensin 5, Thr64-Arg94, Accession # Q01523
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1670
Test Specificity Detects human alpha-Defensin 5 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.