Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-Defensin 5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Defensin alpha 5 |
---|---|
Clone | 972207 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10248SP
![]() |
R&D Systems
MAB10248SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10248100
![]() |
R&D Systems
MAB10248100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for ELISA.Specifications
Defensin alpha 5 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q01523 | |
1670 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
972207 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DEF5, DEF5defensin, alpha 5, preproprotein, DEFA5, defensin 5, Defensin alpha 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728 | |
E. coli-derived recombinant human alpha-Defensin 5, Thr64-Arg94, Accession # Q01523 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human alpha-Defensin 5 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title