Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-Internexin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4844
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4844 Supplier R&D Systems Supplier No. MAB4844
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Internexin Monoclonal specifically detects alpha-Internexin in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alpha-Internexin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 529724
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16352
Gene Alias 66 kDa neurofilament protein, alphaInternexin, alpha-Inx, FLJ18662, FLJ57501, INA, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66, NF-66alpha-internexin, TXBP-1
Gene Symbols INA
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha-Internexin Val230-Glu450 Accession # Q16352
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9118
Test Specificity Detects human alpha-Internexin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) NF-M or rhVimentin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.