Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-L-Iduronidase/IDUA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.50
Specifications
Antigen | alpha-L-Iduronidase/IDUA |
---|---|
Clone | 452619 |
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Description
alpha-L-Iduronidase/IDUA Monoclonal specifically detects alpha-L-Iduronidase/IDUA in Human samples. It is validated for Western Blot.Specifications
alpha-L-Iduronidase/IDUA | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P35475.2 | |
3425 | |
Mouse myeloma cell line NS0-derived recombinant human alpha-L-Iduronidase/IDUA Ala26-Pro653 (Ala26Thr) Accession # P35475.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human alpha-L-Iduronidase/IDUA in direct ELISAs and Western blots. |
452619 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alphaLIduronidase, IDA, IDUA, MPS1, MPSI | |
IDUA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title