Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-Smooth Muscle Actin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$221.50
Specifications
Antigen | alpha-Smooth Muscle Actin |
---|---|
Clone | 1A4 |
Concentration | 0.2 mg/mL |
Dilution | Intracellular Staining by Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IC1420V025
![]() |
R&D Systems
IC1420V025 |
25 Tests |
Each of 1 for $221.50
|
|
|||||
Description
alpha-Smooth Muscle Actin Monoclonal specifically detects alpha-Smooth Muscle Actin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
alpha-Smooth Muscle Actin | |
0.2 mg/mL | |
Flow Cytometry | |
Alexa Fluor 405 | |
Mouse | |
Human | |
AAT6, ACTA2, Actin alpha 2, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, alphaSmooth Muscle Actin, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, MYMY5, sma, smooth muscle actin | |
ACTA2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human alpha-Smooth Muscle Actin in flow cytometry. |
1A4 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
59 | |
alpha-Smooth Muscle Actin N-terminal Synthetic Decapeptide | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title